Learn More
CHIR 99021 trihydrochloride, Tocris Bioscience™
Hydrochloride salt of CHIR 99021 (Cat. No. 4423); selective GSK-3 inhibitor
Marca: Tocris Bioscience 4953/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Hydrochloride salt of CHIR 99021 (Cat. No. 4423), a highly selective GSK-3 inhibitor.Especificaciones
| CHIR 99021 trihydrochloride | |
| C22H21Cl5N8 | |
| chir-99021 trihydrochloride, chir 99021 trihydrochloride, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl-2-pyrimidinyl amino ethyl amino-3-pyridinecarbonitrile trihydrochloride | |
| 574.72 | |
| 574.72 |
| 1782235-14-6 | |
| 50mg | |
| DSFVSCNMMZRCIA-UHFFFAOYSA-N | |
| 78243722 | |
| >99% |