Learn More
Tocris Bioscience™ CGP 60474
Potent cdk inhibitor
Brand: Tocris Bioscience™ 5471/10
This item is not returnable.
View return policy
Description
CGP 60474 is a potent cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 3 - 80, 9.5, 13, 17 - 60, 200 and 220 nM for cdk2, cdk5, cdk9, cdk1, cdk4 and cdk7, respectively). Also inhibits PKCα in the low micromolar range. Displays reversible G1/S cell cycle arrest in U2-OS cells.Specifications
| 3-[[4-[2-[(3-Chlorophenyl)amino]-4-pyrimidinyl]-2-pyridinyl]amino]-1-propanol | |
| 164658-13-3 | |
| 10 mg |