Learn More
CF53, MedChemExpress
Código de producto. 16470947
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16470947 50 mg 50 miligramos
16464658 10 mg 10 miligramos
16437377 5 mg 5 miligramos
16435496 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16470947 Proveedor Medchem Express N.º de proveedor HY112610
MedChemExpress CF53 is a highly potent, selective and orally active inhibitor of BET protein, with a Ki of Kd of 2.2 nM and an IC50 of 2 nM for BRD4 BD1. CF53 binds to both the BD1 and BD2 domains of BRD2, BRD3, BRD4, and BRDT BET proteins with high affinities, very selective over non-BET bromodomain-containing proteins. CF53 shows potent anti-tumor activity both in vitro and in vivo.
CF53 is a highly potent, selective and orally active inhibitor of BET protein, with a Ki of Kd of 2.2 nM and an IC50 of 2 nM for BRD4 BD1. CF53 binds to both the BD1 and BD2 domains of BRD2, BRD3, BRD4, and BRDT BET proteins with high affinities, very selective over non-BET bromodomain-containing proteins. CF53 shows potent anti-tumor activity both in vitro and in vivo.
Especificaciones
Nombre del producto químico o material CF53
CAS 1808160-52-2
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C24H25N7O2
Cantidad 50 mg
Información de solubilidad DMSO : 110 mg/mL (248.03 mM; Need ultrasonic)
SMILES COC1=C(C2=C(C)ON=C2C)C=C(NC3=C4C(NC5=CC(C6CC6)=NN5C)=NC(C)=N3)C4=C1
Peso molecular (g/mol) 443.5
Formula Weight (peso de la fórmula) 443.5
Porcentaje de pureza 98.09%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death