Learn More
Cloruro de cetalconio, TRC
Product Code. 30134957
Change view
Click to view available options
Cantidad:
100 mg
1 g
10 g
Unit Size:
1 g
10 g
100 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad Unit Size
30134957 10 g 10 g
30004763 1 g 1 g
17994715 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134957 Supplier Toronto Research Chemicals Supplier No. TRCC28050010G
 Artículo restringido
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 122-18-9
Fórmula molecular C25 H46 N . Cl
Peso molecular (g/mol) 396.09
Sinónimo Ammonium, benzylhexadecyldimethyl-, chloride (8CI), Benzenemethanaminium, N-hexadecyl-N,N-dimethyl-, chloride (9CI), Benzylhexadecyldimethylammonium chloride (6CI), Acetoquate CDAC, Acinol, Acquat CDAC, Ammonyx G, Ammonyx T, Baktonium, Banicol, Benzaletas, Benzyl-n-hexadecyldimethylammonium chloride, Benzylcetyldimethylammonium chloride, Benzyldimethylcetylammonium chloride, Benzyldimethylhexadecylammonium chloride, Bicetonium, Bonjela, CDBAC, Cetalkonium chloride, Cetol, Cetyl zephiran, Cetylbenzyldimethylammonium chloride, Cetyldimethylbenzylammonium chloride, Cetylon, DMCBAC, Dehyquart CBB, Dehyquart CDB, Dimethylbenzylcetylammonium chloride, Dimethylbenzylhexadecylammonium chloride, HBAC, Hexadecylbenzyldimethylammonium chloride, Hexadecyldimethylbenzylammonium chloride, N-Benzyl-N-cetyldimethylammonium chloride, N-Hexadecyl-N,N-dimethyl-N-benzylammonium chloride, N-Hexadecyl-N-benzyl-N,N-dimethylammonium chloride, Pharycidin Concentrate, Querton 16BCL, Sanisol C, Spilan, Tetraseptan, Win 357, Zettyn, Zettyn chloride, n-Hexadecyldimethylbenzylammonium chloride
Nombre IUPAC benzil-hexadecil-dimetilazanio; cloruro
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
Specifications
Nombre del producto químico o material Benzyldimethylhexadecylammonium Chloride
Notas de grado de pureza HPLC
Cantidad 10 g
Fórmula InChI InChI=1S/C25H46N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-20-23-26(2,3)24-25-21-18-17-19-22-25;/h17-19,21-22H,4-16,20,23-24H2,1-3H3;1H/q+1;/p-1
Formula Weight (peso de la fórmula) 395.3319
Porcentaje de pureza >95%