Learn More
Tocris Bioscience™ Cerestat
Potent and noncompetitive NMDA antagonist
Marca: Tocris Bioscience™ 4492/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cerestat is a potent and noncompetitive NMDA receptor antagonist (IC50 = 36 nM). Exhibits >70-fold selectivity for NMDA receptor over σ receptor. Neuroprotective in a rat model of focal ischemia.Especificaciones
| Cerestat | |
| N-(3-Ethylphenyl)-N-methyl-N'-1-naphthalenylguanidine hydrochloride | |
| Soluble to 50mM in water and to 100mM in DMSO | |
| Bioactive Small Molecule |
| 137160-11-3 | |
| 50 mg | |
| Solid |
Título del producto
Research Use Only. Not For Human or Veterinary Use