Learn More
Tocris Bioscience™ CEP 1347
Descripción
CEP 1347 is a potent inhibitor of mixed lineage kinases MLK3, MLK1 and MLK2 (IC50 values in vitro are 23-39 nM, 38-61 nM and 51-82 nM, respectively), and blocks downstream c-jun N-terminal kinase (JNK) activation (IC50 = 20 nM). CEP 1347 rescues motor neurons undergoing apoptosis (EC50 = 20 nM), blocks Aβ-induced cortical neuron apoptosis (EC50 ∼51 nM) and, in an animal model of Huntington's disease, restores BDNF mRNA to wild type levels. CEP 1347 is orally bioavailable and brain penetrant.
Especificaciones
Especificaciones
| Nombre del producto químico o material | CEP 1347 |
| CAS | 156177-65-0 |
| Cantidad | 1 mg |
| Fórmula molecular | C33H33N3O5S2 |
| Formula Weight (peso de la fórmula) | 615.76 |
| Peso molecular | 615.763 |