Learn More
Tocris Bioscience™ CDN 1163
SERCA2 allosteric activator
Marca: Tocris Bioscience™ 5869/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CDN 1163 is a SERCA2 allosteric activator. Increases Ca2+-ATPase activity and Ca2+ uptake by ER microsomes from obese mice. Rescues HEK cells from ER stress-induced cell death. Reduces fasting glucose levels and adipose tissue weight, and increases energy expenditure in ob/ob mice. Also improves glucose tolerance and reverses hepatic steatosis in ob/ob mice. Neuroprotective and improves memory and cognition in a mouse Alzheimer's disease model. Orally bioavailable.Especificaciones
| 4-(1-Methylethoxy)-N-(2-methyl-8-quinolinyl)benzamide | |
| 892711-75-0 | |
| 50 mg |