Learn More
Tocris Bioscience™ CDK8/19i
Descripción
CDK8/19i is a potent and selective CDK8 and CDK19 inhibitor (IC50 values are 2.9 and 14.1 nM, respectively). CDK8/19i maintains pluripotency of mouse PSCs in culture. It induces the formation of dome-shaped colonies and upregulates Nanog-GFP reporter and endogenous Nanog expression in mouse ESCs in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-Propyl-3-(4-pyridinyl)imidazo[1,2-b]pyridazin-6-amine |
| CAS | 923596-52-5 |
| Cantidad | 10 mg |
| Objetivo | Cyclin-dependent Kinase Inhibitors |
| Fórmula molecular | C14H15N5 |
| Pureza | 0.98 |