missing translation for 'onlineSavingsMsg'
Learn More

CD90 (Thy-1) Mouse, anti-Human, PE, Clone: 5E10, STEMCELL Technologies

Código de producto. p-7217999
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17168761 25 Tests 25 tests
17158761 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17168761 Supplier Stemcell Technologies Supplier No. 60045PE.1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

  • The 5E10 antibody reacts with CD90 (Thy-1), a GPI-linked membrane glycoprotein that is N-glycosylated at two sites, giving rise to 25 - 37 kDa molecules
  • CD90 has roles in signal transduction, cell adhesion and migration, neurite outgrowth, T cell activation, tumor suppression, and inhibition of the proliferation and differentiation of hematopoietic stem cells
  • It is a known ligand of β2 and β3 integrins and upregulates synthesis of fibronectin, osteonectin and thrombospondin
  • CD90 is broadly expressed, being found on human thymocytes, neurons, some glial cells, fibroblasts, activated endothelial cells, some leukemia cell lines and a distinct subset (<1%) of CD3+CD4+ T cells in human peripheral blood
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)

Specifications

Antígeno CD90 (Thy-1)
Aplicaciones Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
Clasificación Monoclonal
Clon 5E10
Conjugado PE
Especie del huésped Mouse
Cantidad 25 Tests
Primario o secundario Primary
Especies diana Baboon, Cynomolgus, Rhesus Monkey, Pig, Human
Isotype IgG1 κ
Tipo de célula Hematopoietic Stem and Progenitor Cells, Mammary Cells, Mesenchymal Stem and Progenitor Cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.