missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 (Thy-1) Mouse, anti-Human, PE, Clone: 5E10, STEMCELL Technologies
Descripción
- The 5E10 antibody reacts with CD90 (Thy-1), a GPI-linked membrane glycoprotein that is N-glycosylated at two sites, giving rise to 25 - 37 kDa molecules
- CD90 has roles in signal transduction, cell adhesion and migration, neurite outgrowth, T cell activation, tumor suppression, and inhibition of the proliferation and differentiation of hematopoietic stem cells
- It is a known ligand of β2 and β3 integrins and upregulates synthesis of fibronectin, osteonectin and thrombospondin
- CD90 is broadly expressed, being found on human thymocytes, neurons, some glial cells, fibroblasts, activated endothelial cells, some leukemia cell lines and a distinct subset (<1%) of CD3+CD4+ T cells in human peripheral blood
- This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Especificaciones
Especificaciones
| Antígeno | CD90 (Thy-1) |
| Aplicaciones | Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 5E10 |
| Conjugado | PE |
| Especie del huésped | Mouse |
| Cantidad | 25 Tests |
| Primario o secundario | Primary |
| Especies diana | Baboon, Cynomolgus, Rhesus Monkey, Pig, Human |
| Isotype | IgG1 κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?