missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Mouse, anti-Human, Unconjugated, Clone: HI9a, STEMCELL Technologies
Descripción
- The HI9a antibody reacts with an extracellular epitope on human CD9, a 24 kDa single-chain type III transmembrane glycoprotein
- CD9 is broadly expressed, including on the surface of monocytes, developing B cells, activated T cells, granulocytes, epithelial cells, endothelial cells, and platelets (mainly the α-granules) and is also a marker characteristic of extracellular vesicles
- CD9 belongs to the tetraspanin family of proteins, which contain four transmembrane domains, two extracellular loops, and short cytoplasmic N- and C-termini
- It is involved in mediating several cellular processes, including angiogenesis, by promoting cell adhesion, migration, and signal transduction, and is involved in cell fusion processes linked to fertilization, osteoclastogenesis, and myogenesis
Especificaciones
Especificaciones
| Antígeno | CD9 |
| Aplicaciones | Flow Cytometry, Western Blot |
| Clasificación | Monoclonal |
| Clon | HI9a |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Cantidad | 100 μg |
| Primario o secundario | Primary |
| Especies diana | African Green Monkey, Baboon, Cynomolgus, Rhesus Monkey, Human |
| Isotype | IgG1 κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?