missing translation for 'onlineSavingsMsg'
Learn More

CD80/B7-1 Mouse anti-Human, Mouse, Porcine, Clone: 1E2F10, Proteintech

Código de producto. 16858255 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16858255 20 μL 20 microlitros
16848255 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16858255 Supplier Proteintech Supplier No. 664061IG20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Involved in the costimulatory signal essential for T lymphocytes activation. T cell proliferation and cytokine production is induced by the binding of CD28 or CTLA4 to this receptor.

Specifications

Antígeno CD80/B7-1
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 1E2F10
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica CD80
N.º de referencia del gen P33681, Q00609
Alias de gen Activation B7 1 antigen, B7, B7 1, B7-1, BB1, CD28LG, CD28LG1, CD80, CD80 molecule, CTLA 4 counter receptor B7.1, LAB7
Símbolos de los genes CD80
Especie del huésped Mouse
Inmunógeno CD80/B7-1 Fusion Protein Ag5615
Método de purificación Protein G
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12519, 397161, 941
Especies diana Pig, Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.