missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 (NCAM) Mouse, anti-Human, PE, Clone: HCD56, STEMCELL Technologies
Descripción
- The HCD56 antibody reacts with CD56 (NCAM), a member of the immunoglobulin (Ig) super family containing 5 Ig-like domains and two fibronectin type-3 domains in its extracellular portion
- Multiple isoforms have been identified, including transmembrane and GPI-anchored forms
- CD56 is subject to several types of post-translational modification, including addition of a polysialic acid moiety to the 5th Ig-like domain
- The ∼140 kDa isoform of CD56 is considered the prototypic cell surface marker on human NK cells
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ HLA Chimerism Buffy Coat CD56 Positive Selection Kit (Catalog #17875) and EasySep™ Human CD56 Positive Selection Kit II (Catalog #17855); partial blocking may be observed, as well as EasySep™ Human CD2 Positive Selection Kit II (Catalog #17883) and EasySep™ Human NK Cell Enrichment Kit (Catalog #19055)
Especificaciones
Especificaciones
| Antígeno | CD56 (NCAM) |
| Aplicaciones | CyTOF, Flow Cytometry, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | HCD56 |
| Conjugado | PE |
| Especie del huésped | Mouse |
| Cantidad | 25 Tests |
| Primario o secundario | Primary |
| Especies diana | Human |
| Isotype | IgG1 κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?