missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49f (Integrin α6) Rat, anti-Mouse, FITC, Clone: GoH3, STEMCELL Technologies
Descripción
- The GoH3 antibody reacts with CD49f (integrin α6), an ∼150 kDa transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) or CD104 (integrin β4) to form the heterodimeric receptors VLA-6 and α6β4, which bind the extracellular matrix protein laminin
- CD49f is a disulfide-linked dimer comprising an ∼120 kDa heavy chain and an ∼30 kDa membrane-bound light chain
- Splice variants exist, which affect the cytoplasmic domain of the protein
- CD49f is expressed on the surface of T cells, monocytes, platelets, placental trophoblasts, epithelial cells, and endothelial cells
- It is involved in cell adhesion and regulating signaling pathways involved in a variety of processes, including the activation and proliferation of T cells, and the differentiation and maintenance of stem cell pluripotency
- CD49f is considered the most important marker for selecting mouse mammary stem and progenitor cells
- The GoH3 antibody reacts with an extracellular epitope on CD49f
Especificaciones
Especificaciones
| Antígeno | CD49f (Integrin α6) |
| Aplicaciones | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Clasificación | Monoclonal |
| Clon | GoH3 |
| Conjugado | FITC |
| Especie del huésped | Rat |
| Cantidad | 100 Tests |
| Primario o secundario | Primary |
| Especies diana | Baboon, Cynomolgus, Pig, Human |
| Isotype | IgG2a κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?