missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies CD437
Beskrivelse
- CD437 is the prototypical adamantyl arotinoid of the retinoid-related molecule family that acts as a selective agonist of retinoic acid receptor (RAR)γ (Kd = 6.5 μM, 2.5 μM, and 77 nM for RARα, β, and γ, respectively; Bernard et al.; Pérez-Rodríguez et al.)
- Increases speed and number of pre-induced pluripotent stem (iPS) cell colonies generated from mouse embryonic fibroblasts (MEFs) transfected with Oct4, Sox2, c-Myc, and Klf4 (Wang et al.)
- Induces cell cycle arrest and apoptosis in a variety of cancer cells (Fontana and Rishi; Jin et al.; Li et al.; Valli et al.)
- Decreases mRNA expression of squamous differentiation markers cytokeratin 1, involucrin, and SPR1 in the human head and neck squamous cell carcinoma cell line, UMSCC22B (Sun et al.)
Tekniske data
Tekniske data
| CAS | 125316-60-1 |
| Sinónimo | AHPN |
| Cantidad | 25 mg |
| Fórmula molecular | C27H26O3 |
| Pureza | ≥95% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?