missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD300g Polyclonal Antibody

Descripción
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CLM-9/TREM4.
CD300g, also known as Immunoglobulin-like Transcripts 4 (ILT4) or CMRF35-like molecule 1 (CLM-1), is a gene located on chromosome 17q25.1, encoding a member of the CD300 family of molecules involved in modulating immune responses. CD300g is expressed on myeloid cells, particularly macrophages and dendritic cells, and plays a role in regulating cell activation and inflammatory processes. It functions as an inhibitory receptor, interacting with phosphatidylserine and other lipid ligands to modulate immune cell activity, influencing cytokine production and phagocytosis. The expression and signaling pathways of CD300g contribute to immune homeostasis and are of particular interest in contexts like autoimmune diseases, infection, and cancer, where immune regulation is disrupted. Research into CD300g aims to uncover potential therapeutic applications in modulating immune functions and offers insights into its role in balancing immune responses in health and disease.
Especificaciones
Especificaciones
| Antígeno | CD300g |
| Aplicaciones | Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | CD300LG |
| N.º de referencia del gen | Q6UXG3 |
| Alias de gen | 2310016B05Rik; CD300 antigen like family member G; CD300 antigen-like family member G; CD300 molecule like family member g; Cd300 molecule-like family member G; CD300g; CD300g antigen; Cd300lg; Clm9; CLM-9; CLM9Trem4; CMRF35-like molecule 9; CMRF-35-like molecule-9; D11Ertd736e; Nepmucin; RGD1311074; TREM4; TREM-4; Triggering receptor expressed on myeloid cells 4; UNQ422/PRO846 |
| Símbolos de los genes | CD300LG |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Spot an opportunity for improvement?