missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD28 Recombinant Mouse Monoclonal Antibody (JJ316)
GREENER_CHOICE

Código de producto. 17971251
Change view
Click to view available options
Cantidad:
200 μg
Tamaño de la unidad:
200 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17971251 200 μg 200 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17971251 Proveedor Invitrogen™ N.º de proveedor MA547868

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Recombinant Monoclonal Antibody

Specificity: Recognises rat CD28 and can trigger T-cell proliferation in vitro and in vivo.

CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary's Disease.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD28
Aplicaciones Flow Cytometry, Functional Assay, Immunohistochemistry, T-Cell Activation, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon JJ316
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.02% ProClin 300
génica Cd28
N.º de referencia del gen P31042
Alias de gen antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44
Símbolos de los genes Cd28
Especie del huésped Mouse
Inmunógeno Made by immunizing BALB/c mice with rat CD28 cDNA-transfected A20/J cells.
Método de purificación Protein A
Cantidad 200 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 25660
Especies diana Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.