missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD200R3 Monoclonal Antibody (Ba103), FITC

Código de producto. 17976471
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17976471 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17976471 Proveedor Invitrogen™ N.º de proveedor MA549007

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

The monoclonal antibody Ba103 recognizes mouse CD200 Receptor-like 3 (CD200R3). Monoclonal antibody Ba103 activates basophils ex vivo and elicits systemic anaphylaxis when administered in vivo. Cross-linking of CD200R3 on MC/9 cells by monoclonal antibody Ba103 induces degranulation. Furthermore, monoclonal antibody Ba103 is useful for depletion of basophils. This antibody does not cross-react with CD200R, CD200R2, CD200R4 or CD200R5.

CD200R3 is a dimeric cell surface receptor also known as CD200RLb. CD200R3 contains two Ig-like extracellular domains and as such is a member of the immunoglobulin superfamily of receptors. The intracellular region of CD200R3 associates with the adaptor molecule DAP12, and possibly also DAP10, CD3 zeta and FcR gamma-chain, indicating that it is capable of transducing stimulatory signals. In this respect CD200R3 bears similarity to activatory receptors CD200R2 and CD200R4. In contrast, an inhibitory receptor CD200R (also known as CD200R1) relies on other adaptors. CD200R3 is primarily expressed by basophils and mast cells, cells known to be involved in anti-parasitic immune responses and frequently also allergic reactions. The surface expression of CD200R3 decreases when basophils are activated by ligating their CD16 and CD32. Remarkably, most of this downregulation is observed within the first 30 minutes after activation. Unlike CD200R, CD200R3 does not bind CD200.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD200R3
Aplicaciones Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon Ba103
Concentración 100 μg/mL
Conjugado FITC
Formulación PBS with 1% BSA and 0.02% sodium azide
génica CD200R3
N.º de referencia del gen Q5UKY4
Alias de gen 4733401I18Rik; 4833409J19Rik; AI505817; BB106877; CD200 cell surface glycoprotein receptor 3; CD200 cell surface glycoprotein receptor-like 3; CD200 cell surface glycoprotein receptor-like b; CD200 receptor 3; CD200 receptor-like 3; Cd200r3; CD200RLb; cell surface glycoprotein CD200 receptor 3; Cell surface glycoprotein OX2 receptor 3; mCD200RLb
Símbolos de los genes CD200R3
Especie del huésped Rat
Inmunógeno Mouse primary basophils; cell-enriched bone marrow cells.
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 74603
Especies diana Mouse
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.