Learn More
CD11a/Integrin Alpha L Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 16894804 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16894804 20 μL 20 microlitros
16884804 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16894804 Proveedor Proteintech N.º de proveedor 155741AP20UL
Rabbit Polyclonal Antibody
The CD11a/Integrin Alpha L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD11a/Integrin Alpha L. This antibody recognizes human antigen. The CD11a/Integrin Alpha L antibody has been validated for the following applications: IHC, ELISA analysis.
Especificaciones
Antígeno CD11a/Integrin Alpha L
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clasificación Polyclonal
Concentración 187 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 3683
Especies diana Human
Isotype IgG