missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCRL2 Polyclonal Antibody

Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey (95%) Marmoset (90%).
CCRL2 (C-C chemokine receptor-like 2), also known as chemokine receptor X, human chemokine receptor (HCR), CRAM-A or CRAM-B, is a seven pass transmembrane protein expressed by monocytes, neutrophils, and dendritic cells of immunal tissues (predominanly spleen, fetal liver, bone marrow and lymph node). It shares over 40% homology with other C-C chemokine receptors but does not share the conserved DRY motif which is among the important motifs necessary for signalling and ligand-binding. C-C chemokine receptors are G-protein coupled, seven pass transmembrane domain proteins whose major physiological role is to function in the chemotaxis of T cells and phagocytic cells to areas of inflammation.
Especificaciones
Especificaciones
| Antígeno | CCRL2 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide |
| génica | CCRL2 |
| N.º de referencia del gen | O00421 |
| Alias de gen | 1810047I05Rik; Ackr5; atypical chemokine receptor 5; C-C chemokine receptor-like 2; C-C motif chemokine receptor like 2; CCR11; CCR6; Ccrl2; chemokine (C-C motif) receptor-like 2; chemokine C-C motif receptor-like 2; chemokine C-C motif receptor-like 2-like; chemokine receptor CCR11; Chemokine receptor X; CKRX; Cmkbr1l2; CRAM; CRAM-A; CRAM-B; E01; G-protein coupled beta chemokine receptor; HCR; Lccr; L-CCR; Lipopolysaccharide-inducible C-C chemokine receptor; LOC100719164; MGC116710; Putative MCP-1 chemokine receptor |
| Símbolos de los genes | CCRL2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?