Learn More
CCR6 inhibitor 1, MedChemExpress
Código de producto. 16465516
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16465516 100 mg 100 miligramos
16447397 5 mg 5 miligramos
16404688 10 mg 10 miligramos
16400977 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16465516 Proveedor Medchem Express N.º de proveedor HY112701
MedChemExpress CCR6 inhibitor 1 is a potent and selective CCR6 inhibitor, with IC50s of 0.45 and 6 nM for monkey and human CCR6, much more selective at CCR6 over human CCR1 (IC50, > 30000 nM), and CCR7 (IC50, 9400 nM). CCR6 inhibitor 1 markedly blocks ERK phosphorylation. CCR6 inhibitor 1 is used in the research of autoimmune diseases and cancer.
CCR6 inhibitor 1 is a potent and selective CCR6 inhibitor, with IC50s of 0.45 and 6 nM for monkey and human CCR6, much more selective at CCR6 over human CCR1 (IC50, > 30000 nM), and CCR7 (IC50, 9400 nM). CCR6 inhibitor 1 markedly blocks ERK phosphorylation. CCR6 inhibitor 1 is used in the research of autoimmune diseases and cancer.
Especificaciones
Nombre del producto químico o material CCR6 inhibitor 1
CAS 2437547-04-9
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C24H23F3N4O3S
Cantidad 100 mg
Información de solubilidad DMSO : ≥ 125 mg/mL (247.76 mM)
SMILES O=S([C@H](CC1)CC[C@@H]1NC2=NC=C(C(F)(F)F)C=C2)(C3=CC=C(C4=CC=NC(C(N)=O)=C4)C=C3)=O
Peso molecular (g/mol) 504.52
Formula Weight (peso de la fórmula) 504.52
Porcentaje de pureza 99.87%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease