missing translation for 'onlineSavingsMsg'
Learn More

CCG-203971, MedChemExpress

Código de producto. 16452986
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16452986 25 mg 25 miligramos
16475146 100 mg 100 miligramos
16474188 10 mg 10 miligramos
16460537 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452986 Supplier Medchem Express Supplier No. HY108361

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress CCG-203971 is a second-generation Rho/MRTF/SRF pathway inhibitor. CCG-203971 potently targets RhoA/C-activated SRE-luciferase (IC50 =6.4 μM). CCG-203971 inhibits PC-3 cell migration with an IC50 of 4.2 μM. Potential anti-metastasis Agent.

CCG-203971 is a second-generation Rho/MRTF/SRF pathway inhibitor. CCG-203971 potently targets RhoA/C-activated SRE-luciferase (IC50 =6.4 μM). CCG-203971 inhibits PC-3 cell migration with an IC50 of 4.2 μM. Potential anti-metastasis Agent.

Specifications

Nombre del producto químico o material CCG-203971
CAS 1443437-74-8
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C23H21ClN2O3
Cantidad 25 mg
Información de solubilidad DMSO : 250 mg/mL (611.43 mM; Need ultrasonic)
SMILES O=C(C1CN(C(C2=CC=CC(C3=CC=CO3)=C2)=O)CCC1)NC4=CC=C(Cl)C=C4
Peso molecular (g/mol) 408.88
Formula Weight (peso de la fórmula) 408.88
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.