Learn More
Catalpol, MedChemExpress
Código de producto. 16464897
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16464897 50 mg 50 miligramos
16449118 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16464897 Proveedor Medchem Express N.º de proveedor HYN0820
MedChemExpress Catalpol (Catalpinoside), an iridoid glycoside found in Rehmannia glutinosa. Catalpol has neuroprotective, hypoglycemic, anti-inflammatory, anti-cancer, anti-spasmodic, anti-oxidant effects and anti-HBV effects.
Catalpol (Catalpinoside), an iridoid glycoside found in Rehmannia glutinosa. Catalpol has neuroprotective, hypoglycemic, anti-inflammatory, anti-cancer, anti-spasmodic, anti-oxidant effects and anti-HBV effects.
Especificaciones
Nombre del producto químico o material Catalpol
CAS 2415-24-9
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C15H22O10
Cantidad 50 mg
Sinónimo Catalpinoside
Información de solubilidad DMSO : ≥ 30 mg/mL (82.80 mM)
SMILES OC[C@@]12[C@]([C@@H]3O[C@]([C@@H]([C@@H](O)[C@@H]4O)O)([H])O[C@@H]4CO)([H])[C@](C=CO3)([H])[C@H](O)[C@@H]1O2
Peso molecular (g/mol) 362.33
Formula Weight (peso de la fórmula) 362.33
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid