missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Caspase 14 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13239448
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
13239448 100 μL 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 13239448 Proveedor Invitrogen™ N.º de proveedor PA522194

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Recommended positive controls: A431, PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Caspase 14 encodes a member of the cysteine-aspartic acid protease (caspase) family, which plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes that undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This caspase has been shown to be processed and activated by caspase 8 and caspase 10 in vitro, and by anti-Fas agonist antibody or TNF-related apoptosis inducing ligand in vivo. The expression and processing of this caspase may be involved in keratinocyte terminal differentiation, which is important for the formation of the skin barrier. Diseases associated with CASP14 include Ichthyosis, Congenital, Autosomal Recessive 12 and Autosomal Recessive Congenital Ichthyosis.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Caspase 14
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
génica Casp14
N.º de referencia del gen P31944
Alias de gen apoptosis-related cysteine protease; CASP14; CASP-14; caspase 14; caspase 14, apoptosis-related cysteine peptidase; caspase 14, apoptosis-related cysteine protease; Caspase14; caspase-14; Caspase-14 subunit p10, mature form; Caspase-14 subunit p17, mature form; Caspase-14 subunit p20, intermediate form; Caspase-14 subunit p8, intermediate form; caspase-14; LOW QUALITY PROTEIN: caspase-14; MICE; Mini-ICE
Símbolos de los genes Casp14
Especie del huésped Rabbit
Inmunógeno Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Caspase 14. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23581
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.