Learn More
Carglumic Acid, MedChemExpress
Código de producto. 16482259
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
200 mg
500 mg
1 g
Tamaño de la unidad:
1 g
200 miligramos
500 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16482259 1 g 1 g
16471436 200 mg 200 miligramos
16492196 500 mg 500 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16482259 Proveedor Medchem Express N.º de proveedor HYB0711
MedChemExpress Carglumic acid (N-Carbamyl-L-glutamic acid), a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator, is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency.
Carglumic acid (N-Carbamyl-L-glutamic acid), a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator, is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency.
Especificaciones
Nombre del producto químico o material Carglumic Acid
Forma física Solid
CAS 1188-38-1
Color Blanco
Cantidad 1 g
Notas de grado de pureza Research
Fórmula molecular C6H10N2O5
Para utilizar con (aplicación) Cancer-programmed cell death
Sinónimo N-Carbamyl-L-glutamic acid
Información de solubilidad DMSO : 100 mg/mL (525.90 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (43.81 mM; Need ultrasonic)
SMILES O=C(O)CC[C@@H](C(O)=O)NC(N)=O
Peso molecular (g/mol) 190.15
Formula Weight (peso de la fórmula) 190.15
Porcentaje de pureza 98.0%