missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CARD14 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17721864
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17721864 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17721864 Proveedor Invitrogen™ N.º de proveedor PA5120462

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, HeLa. The target is usually found in the following locations: Cytoplasm, Cytoplasm. Immunogen sequence: SLSLHYSNAL QEKELAASRC RSLQEELYLL KQELQRANMV SSCELELQEQ SLRTASDQES GDEELNRLKE ENEKLRSLTF SLAEKDILEQ SLDEARGSRQ ELVERIHSLR ERAVAAERQR EQYWEEKEQT LLQFQKSKMA CQLYREKVNA LQAQVCELQK ERDQAYSARD SAQREISQSL VEKDSLRRQV FELTDQVCEL RTQLRQLQAE PPGVLKQEAR TREPCPREKQ RLVRMHAICP RDDSDCSLVS STESQLLSDL SATSSRELVD SFRSSSPAPP SQQSLYKRVA EDFGEEPWSF S.

Membrane-associated guanylate kinase (MAGUK) family members localize to the plasma membrane and function as molecular scaffolds for the assembly of multi-protein complexes. The MAGUK family includes several mammalian proteins related to the Drosophila tumor suppressor discs-large (dlg) gene product, such as postsynaptic proteins, GKAPs, the tight junction associated proteins (ZO-1-3) and the caspase-associated recruitment domain (CARD) proteins: CARD 6, CARD 8-12 and CARD 14. CARD 14 is a 1,004 amino acid protein consisting of an N-terminal CARD domain, a central coiled-coil domain and a C-terminal tripartite domain comprised of a PDZ domain, an Src homology 3 domain and a GUK domain with homology to guanylate kinase. CARD 14 is expressed in the placenta where it positively regulates apoptosis. CARD 14 also controls NF-kappaB activation by phosphorylating BCL10, a signaling protein that activates NF-kappaB through the I kappaB kinase complex. Epigallocatechin-3-gallate (EGCG) is a polyphenol that induces the expression of CARD 14.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CARD14
Aplicaciones ELISA, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 2.12 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
génica CARD14
N.º de referencia del gen Q99KF0, Q9BXL6
Alias de gen Bcl10-interacting MAGUK protein 2; BIMP2; Card14; CARD-containing MAGUK protein 2; Carma 2; CARMA2; caspase recruitment domain family member 14; caspase recruitment domain family, member 14; caspase recruitment domain-containing protein 14; PRP; PSORS2; PSS1
Símbolos de los genes CARD14
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human CARD14 (NP_001244899.1).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 170720, 688292, 79092
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.