Learn More
Carboxifosfamida-d4, TRC
Product Code. 30365334
Change view
Click to view available options
Cantidad:
10 mg
Unit Size:
10 miligramos
Product Code. Cantidad unitSize
30365334 10 mg 10 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30365334 Supplier Toronto Research Chemicals Supplier No. TRCC18135210MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Carboxyphosphamide-d4
CAS 1246817-74-2
Fórmula molecular C7H11D4Cl2N2O4P
Cantidad 10 mg
Sinónimo 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid-d4, N,N-Bis(2-chloroethyl)phosphorodiamidate Hydracrylic Acid-d4, Asta 5754-d4, Carboxycyclophosphamide-d4, NSC 145124-d4
Fórmula InChI InChI=1S/C7H15Cl2N2O4P/c8-2-4-11(5-3-9)16(10,14)15-6-1-7(12)13/h1-6H2,(H2,10,14)(H,12,13)
SMILES O=P(N(CC([2H])([2H])Cl)CC([2H])([2H])Cl)(N)OCCC(O)=O
Nombre IUPAC Ácido 3-[amino-[bis(2-cloroetil)amino]fosforil]oxipropanoico
Peso molecular (g/mol) 297.11
Formula Weight (peso de la fórmula) 296.0398