missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPG Polyclonal Antibody

Descripción
This antibody is predicted to react with dog based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in non-muscle cells. Alternatively spliced transcript variants have been observed, but have not been fully described.
Especificaciones
Especificaciones
| Antígeno | CAPG |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| génica | CAPG |
| N.º de referencia del gen | P40121 |
| Alias de gen | Actin regulatory protein CAP-G; actin-capping protein GCAP39; actin-regulatory protein CAP-G; AFCP; Capg; capping actin protein, gelsolin like; capping protein (actin filament), gelsolin-like; epididymis secretory protein Li 66; gCap39; gelsolin-like capping protein; HEL-S-66; macrophage capping protein; macrophage-capping protein; mbh1; MCP; myc basic motif homolog 1 |
| Símbolos de los genes | CAPG |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?