Learn More
Canrenone, 98%, Thermo Scientific™
Inhibits aldosterone biosynthesis and blocks ouabain effects
Marca: Thermo Scientific Alfa Aesar J60238.06
| Cantidad | 5g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| C22H28O3 | |
| MFCD00071735 | |
| canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
| (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione |
Tekniske data
| Canrenone | |
| C22H28O3 | |
| 5g | |
| canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
| CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C | |
| 340.463 | |
| 340.46 |
| 976-71-6 | |
| MFCD00071735 | |
| 14,1752 | |
| UJVLDDZCTMKXJK-WNHSNXHDSA-N | |
| (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione | |
| 13789 | |
| 98% |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.