missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CaMKIV Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15877213
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15877213 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15877213 Proveedor Invitrogen™ N.º de proveedor PA568804

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%.

CAMK4 (CaMKIV) is a serine/threonine protein kinase that phosphoryates CREB, oncoprotein 18, and SRF. It has limited tissue distribution, and has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells. The nuclear localization of this protein is consistent with its role in mediating calcium-dependent gene expression. CaMKIV is particularly abundant in testis, T-cells, and neurons but is also found in other tissues to varying degrees. In neurons, CaMKIV is thought to play an important role in synaptic plasticity via its gene regulatory effects. In T-cells, this protein plays an important role in calcium signaling which could affect the transcription regulatory protein, nuclear factor of activated T-cells (NFAT). CaMKIV is encoded, along with calspermin, by the CaMKIV gene. It has been found that, in testes, CaMKIV is expressed in germ cells and found to be associated with chromatin. The association of CaMKIV with chromatin suggests a potential role in chromatin remodeling during nuclear condensation in spermatids.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CaMKIV
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica CAMK4
N.º de referencia del gen P08414
Alias de gen A430110E23Rik; AI666733; brain Ca(2+)-calmodulin-dependent protein kinase type IV; brain Ca++-calmodulin-dependent protein kinase type IV; Ca2+/calmodulin-dependent protein kinase type IV/Gr; Calcium/calmoduli; calcium/calmodulin dependent protein kinase IV; calcium/calmodulin-dependent protein kinase IV; calcium/calmodulin-dependent protein kinase IV L homeolog; calcium/calmodulin-dependent protein kinase type IV; calcium/calmodulin-dependent protein kinase type IV catalytic chain; Calmodulin-dependent protein kinase IV; calspermin; CAM kinase- GR; CAM kinase IV; CAM kinase-GR; CAMK; CaMK IV; camk4; camk4.L; camk4-A; CAMK-GR; CAMKIV; CaMKIV/Gr; Ccdpk; D18Bwg0362e; EC 2.7.11.17; KCC4; kinase CaMK4; OTTHUMP00000222818; RATCCDPK; xCaM; XELAEV_18008011mg
Símbolos de los genes CAMK4
Especie del huésped Rabbit
Inmunógeno synthetic peptide directed towards the C-terminal region of mouse CaMKIV.
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12326
Especies diana Mouse
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.