Learn More
Cloruro de camfetamina, TRC
Product Code. 30180375
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
1 mg
5 mg
missing translation for 'unitSize'
1 miligramo
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30180375 1 mg 1 miligramo
30179822 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30180375 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC1500501MG
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39628-11-0
Fórmula molecular C14 H19 N . Cl H
Peso molecular (g/mol) 237.77
Sinónimo Camfetamine Hydrochloride, N-Methyl-3-phenylnorbornan-2-amine Hydrochloride
Nombre IUPAC N-metil-2-fenilbiciclo[2.2.1]heptano-3-amina; Cloruro
SMILES Cl.CNC1C2CCC(C2)C1c3ccccc3
Specifications
Nombre del producto químico o material Camfetamine Hydrochloride (N-Methyl-3-phenylnorbornan-2-amine Hydrochloride)
Cantidad 1 mg
Fórmula InChI InChI=1S/C14H19N.ClH/c1-15-14-12-8-7-11(9-12)13(14)10-5-3-2-4-6-10;/h2-6,11-15H,7-9H2,1H3;1H
Formula Weight (peso de la fórmula) 237.1284