missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calponin 1 Monoclonal Antibody (CALP)
GREENER_CHOICE

Código de producto. 11374483
Cambiar vista
Click to view available options
Cantidad:
500 μL
Tamaño de la unidad:
500 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11374483 500 μL 500 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11374483 Proveedor Invitrogen™ N.º de proveedor MA511620

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA5-11620 targets Calponin-1 in IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-11620 immunogen is crude human uterus extract.

Calponin, a thin filament associated protein is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34kDa and 29kDa. Expression of the 29kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Calponin 1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon CALP
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
génica CNN1
N.º de referencia del gen P51911, Q08091, Q08290
Alias de gen Basic calponin; calponin; calponin 1; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; calponin H1, smooth muscle; Calponin-1; calponins, basic; CN; CNN1; CnnI; epididymis secretory protein Li 14; h1-calponin; HEL-S-14; similar to calponin 1, basic, smooth muscle; Sm-Calp; SMCC; smooth muscle calponin h1
Símbolos de los genes CNN1
Especie del huésped Mouse
Inmunógeno Crude human uterus extract.
Método de purificación Protein G
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1264, 12797, 65204
Especies diana Human, Mouse, Rat
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.