missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calmyrin Polyclonal Antibody
GREENER_CHOICE

Código de producto. 16304535 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16304535 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16304535 Proveedor Invitrogen™ N.º de proveedor PA595262

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Human CIB1 shares 93.5% amino acid (aa) sequence identity with both mouse and rat CIB1. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A431 whole cell, rat PC-12 whole cell, mouse brain tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alphabeta, which may implicate this protein as a regulatory molecule for alphabeta.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Calmyrin
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 500 μg/mL
Conjugado Unconjugated
Formulación PBS with 5mg BSA and 0.05mg sodium azide
génica Cib1
N.º de referencia del gen Q99828, Q9R010, Q9Z0F4
Alias de gen calcium and integrin binding 1; calcium and integrin binding 1 (calmyrin); calcium and integrin binding protein 1; calcium and integrin binding, protein kinase interacting protein; Calcium- and integrin-binding protein; calcium and integrin-binding protein 1; calmyrin; CIB; CIB1; Cibkip; CIBP; DNA-dependent protein kinase interacting protein; DNA-PK interaction protein; DNA-PKcs-interacting protein; Kinase-interacting protein; KIP; KIP1; LOW QUALITY PROTEIN: calcium and integrin-binding protein 1; PRKDCIP; protein kinase, DNA activated, catalytic polypeptide interacting protein; Sip2-28; SNK-interacting protein 2-28; testicular secretory protein Li 9
Símbolos de los genes Cib1
Especie del huésped Rabbit
Inmunógeno E.coli-derived human CIB1 recombinant protein (Position: S6-L191).
Método de purificación Affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10519, 23991, 81823
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.