Learn More
Calcifediol-d6, TRC
Product Code. 30343958
Change view
Click to view available options
Cantidad:
0.5 mg
Unit Size:
0.50 miligramo
Product Code. Cantidad unitSize
30343958 0.5 mg 0.50 miligramo
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30343958 Supplier Toronto Research Chemicals Supplier No. TRCC1257020.5MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material D6-25-Hydroxyvitamin D3
CAS 78782-98-6
Fórmula molecular C27 2H6 H38 O2
Cantidad 0.5 mg
Sinónimo 1H-Indene-1-pentanol, 4-[(2Z)-2-[(5S)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]octahydro-ε,7a-dimethyl-α,α-bis(methyl-d3)-, (εR,1R,3aS,4E,7aR)-, 9,10-Secocholesta-5,7,10(19)-triene-26,26,26,27,27,27-d6-3,25-diol, (3β,5Z,7E)- (9CI), (εR,1R,3aS,4E,7aR)-4-[(2Z)-2-[(5S)-5-Hydroxy-2-methylenecyclohexylidene]ethylidene]octahydro-ε,7a-dimethyl-α,α-bis(methyl-d3)-1H-indene-1-pentanol, 25-Hydroxyvitamin D3-(26,26,26,27,27,27-d6), 26,26,26,27,27,27-Hexadeutero-25-hydroxyvitamin D3, 25-Hydroxycholecalciferol-26,26,26,27,27,27-D6, 25-Hydroxyvitamin D3-d6, D6-25-Hydroxyvitamin D3 (26,26,26,27,27,27-D6), Calcifediol-d6, Vitamin D3-25-hydroxy d6 (26,26,26,27,27,27-d6)
Fórmula InChI InChI=1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1/i3D3,4D3
SMILES [2H]C([2H])([2H])C(O)(CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C)C([2H])([2H])[2H]
Nombre IUPAC (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-metil-1-[(2R)-7,7,7-trideuterio-6-hidroxi-6-(trideuteriometil)heptan-2-il]-2,3,3a,5,6,7-hexahidro-1H-inden-4-ilideno]etilideno]-4-metilidenociclohexan-1-ol
Peso molecular (g/mol) 406.67
Formula Weight (peso de la fórmula) 406.3718