Learn More
CA-074 methyl ester, MedChemExpress
Código de producto. 16485697
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
25 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
25 miligramos
5 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16485697 5 mg 5 miligramos
16462958 10 mg 10 miligramos
16452556 25 mg 25 miligramos
16420639 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16485697 Proveedor Medchem Express N.º de proveedor HY100350
MedChemExpress CA-074 methyl ester is a specific inhibitor of Cathepsin B, which has potent bioactivities such as neuroprotective, anti-cancer, and anti-inflamatory effects.
CA-074 methyl ester is a specific inhibitor of Cathepsin B, which has potent bioactivities such as neuroprotective, anti-cancer, and anti-inflamatory effects.
Especificaciones
Nombre del producto químico o material CA-074 methyl ester
CAS 147859-80-1
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C19H31N3O6
Cantidad 5 mg
Sinónimo CA-074Me
Información de solubilidad DMSO : 100 mg/mL (251.59 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C([C@@H]1[C@@H](C(NCCC)=O)O1)N[C@]([C@@H](C)CC)([H])C(N(CCC2)[C@@H]2C(OC)=O)=O
Peso molecular (g/mol) 397.47
Formula Weight (peso de la fórmula) 397.47
Porcentaje de pureza 98.2%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Cancer-Kinase/protease