Learn More
Tocris Bioscience™ C25-140
Descripción
C25-140 binds directly to TNF receptor-associated factor 6 (TRAF6) and inhibits its interaction with ubiquitin-conjugating enzyme Ubc13. C25-140 reduces TRAF6-Ubc13 activity and decreases NF-κB activation and inflammatory signaling in vitro. C25-140 exhibits selectivity for TRAF6 over other E3 ligases except cIAP1. In in vivo mouse models of autoimmune psoriasis and rheumatoid arthritis, the compound improves disease symptoms. The compound also suppresses the pro-inflammatory signaling activity of IL-17A and prevents its inhibitory effect on the anti-HBV action of IFN-α HepG2 cells.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3-[3,5-Dimethyl-1-(3-methyl-1,2,4-triazolo[4,3-b]pyridazin-6-yl)-1H-pyrazol-4-yl]-1-[4-(phenylmethyl)-1-piperidinyl]-1-propanone |
| CAS | 1358099-18-9 |
| Cantidad | 10 mg |
| Objetivo | Ubiquitin Ligase (E3) Inhibitors |
| Fórmula molecular | C26H31N7O |