Learn More
c-di-AMP, MedChemExpress
Descripción
c-di-AMP (Cyclic diadenylate) is also a bacterial second messenger, which regulates cell growth, survival, and virulence, primarily within Gram-positive bacteria, and also regulates host immune response. c-di-AMP (Cyclic diadenylate) acts as a potent mucosal adjuvant stimulating both humoral and cellular responses.
Especificaciones
Especificaciones
| Nombre del producto químico o material | c-di-AMP |
| CAS | 54447-84-6 |
| Color | Blanco |
| Notas de grado de pureza | Research |
| Fórmula molecular | C20H24N10O12P2 |
| Cantidad | 1 mg |
| Sinónimo | Cyclic diadenylate Cyclic-di-AMP |
| Información de solubilidad | H2O : 3.33 mg/mL (5.06 mM; Need ultrasonic) |
| SMILES | O[C@H]([C@@](OP(O)(OC[C@](O[C@@H](N1C2=NC=NC(N)=C2N=C1)[C@@H]3O)([H])[C@@]3([H])O4)=O)([H])[C@](COP4(O)=O)([H])O5)[C@@H]5N6C7=NC=NC(N)=C7N=C6 |
| Peso molecular (g/mol) | 658.41 |
| Mostrar más |