Learn More
C 646, Tocris Bioscience™
Código de producto. 18075432
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18075432 50 mg 50 miligramos
18004992 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18075432 Proveedor Tocris Bioscience N.º de proveedor 4200/50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Selective p300/CBP inhibitor
Selective p300/CREB-binding protein (CBP) inhibitor (Ki = 400 nM). Selective for p300 over six other histone acetyltransferases (HATs). Suppresses histone H3 and H4 acetylation in mouse fibroblast cell lines.
This compound is a mixture of Z:E isomers (∽ 80:20).
TRUSTED_SUSTAINABILITY
CAS 328968-36-1
Fórmula molecular C24H19N3O6
Peso molecular (g/mol) 445.43
Número MDL MFCD01784780
Clave InChI HEKJYZZSCQBJGB-UHFFFAOYSA-N
Sinónimo 4-4-5-4,5-dimethyl-2-nitrophenyl furan-2-yl methylidene-3-methyl-5-oxopyrazol-1-yl benzoic acid
PubChem CID 2871948
Nombre IUPAC 4-(4-{[5-(4,5-dimethyl-2-nitrophenyl)furan-2-yl]methylidene}-3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid
SMILES CC1=NN(C(=O)C1=CC1=CC=C(O1)C1=CC(C)=C(C)C=C1[N+]([O-])=O)C1=CC=C(C=C1)C(O)=O
Especificaciones
Nombre del producto químico o material C 646
Cantidad 50 mg
Formula Weight (peso de la fórmula) 445.42
Porcentaje de pureza >98%