Learn More
BX-912, MedChemExpress
Código de producto. 16430677
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
50 mg
100 mg
Tamaño de la unidad:
100 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16430677 50 mg 50 miligramos
16417067 5 mg 5 miligramos
16405266 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16430677 Proveedor Medchem Express N.º de proveedor HY11005
MedChemExpress BX-912 is a direct, selective, and ATP-competitive PDK1 inhibitor (IC50=26 nM). BX-912 blocks PDK1/Akt signaling in tumor cells and inhibits the anchorage-dependent growth of a variety of tumor cell lines in culture or induces apoptosis.
BX-912 is a direct, selective, and ATP-competitive PDK1 inhibitor (IC50=26 nM). BX-912 blocks PDK1/Akt signaling in tumor cells and inhibits the anchorage-dependent growth of a variety of tumor cell lines in culture or induces apoptosis.
Especificaciones
Nombre del producto químico o material BX-912
Forma física Solid
CAS 702674-56-4
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C20H23BrN8O
Para utilizar con (aplicación) Cancer-Kinase/protease
Información de solubilidad DMSO : ≥ 100 mg/mL (212.16 mM)
SMILES BrC1=C(NCCC2=CNC=N2)N=C(NC3=CC(NC(N4CCCC4)=O)=CC=C3)N=C1
Peso molecular (g/mol) 471.35
Formula Weight (peso de la fórmula) 471.35
Porcentaje de pureza 99.53%