Learn More
Tocris Bioscience™ BX 430
Selective P2X4 allosteric antagonist
Marca: Tocris Bioscience™ 5545/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BX 430 is a selective P2X4 allosteric antagonist (IC50 = 0.54 μM). Exhibits >10-fold selectivity for human P2X4 receptors over P2X1, P2X2, P2X3, P2X5 and P2X7 receptors. Binds noncompetitively at extracellular allosteric site. Effective at human and zebrafish, but not mouse and rat P2X4 receptors.Especificaciones
| N-[2,6-Dibromo-4-(1-methylethyl)phenyl]-N′-(3-pyridinyl)urea | |
| 688309-70-8 | |
| 50 mg |