Learn More
STEMCELL Technologies Butein
Descripción
- Butein is a plant polyphenol, tetrahydroxychalcone that inhibits nuclear factor (NF)-κB (Yang et al; Pandey et al. 2007)
- Butein has been shown to prevent phosphorylation and degradation of TNF-dependent IκBα, an inhibitory subunit of NF-κB (IC50 = 38 μM; Orlikova et al.) Butein also inhibits 5-lipoxygenase (IC50 = 0.01 μM; Sogawa et al.), the enoyl-acyl-carrier protein reductase of Plasmodium falciparum (Ki = 2.97 μM; Sharma et al.), angiotensin-converting enzyme (IC50 = 0.73 mM; Bonesi et al.), and SRC kinase (Pandey et al. 2009)
Especificaciones
Especificaciones
| CAS | 487-52-5 |
| Sinónimo | 2',3,4,4'-Tetrahydroxychalcone |
| Cantidad | 10 mg |
| Fórmula molecular | C15H12O5 |
| Pureza | ≥95% |