Learn More
Buserelin Acetate, TRC
Código de producto. 30006380
Cambiar vista
Click to view available options
Cantidad:
1 mg
10 mg
50 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30006380 10 mg 10 miligramos
30006429 1 mg 1 miligramo
30004307 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30006380 Proveedor Toronto Research Chemicals N.º de proveedor TRCB68974010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
CAS 68630-75-1
Sinónimo 6-[O-(1,1-dimethylethyl)-D-serine]-9-(N-ethyl-L-prolinamide)-1-9-Luteinizing hormone-releasing factor (swine) acetate (1:1)∣1-9-Luteinizing hormone-releasing factor (swine), 6-[O-(1,1-dimethylethyl)-D-serine]-9-(N-ethyl-L-prolinamide)-, monoacetate (salt) (9CI)∣Luteinizing hormone-releasing factor (pig), 6-[O-(1,1-dimethylethyl)-D-serine]-9-(N-ethyl-L-prolinamide)-10-deglycinamide-, monoacetate (salt)∣1-9-Luteinizing hormone-releasing factor (pig), 6-[O-(1,1-dimethylethyl)-D-serine]-9-(N-ethyl-L-prolinamide)-, monoacetate (salt)∣Buserelin acetate∣Conceptal∣Estomal∣Gonaxal∣Sprecur MP∣Suprafact∣Suprecur∣Supremon
Especificaciones
Formato Neat
Fórmula molecular C62H90N16O15
Número MDL MFCD01716229
Peso molecular 1299.48
Clave InChI PYMDEDHDQYLBRT-DRIHCAFSSA-N
SMILES CC(O)=O.CCNC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCN=C(N)N)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](COC(C)(C)C)NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@H](CO)NC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)[C@H](CC1=CN=CN1)NC(=O)[C@@H]1CCC(=O)N1
Nombre IUPAC (2S)-1-[(2S)-2-[(2S)-2-[(2R)-3-(tert-butoxy)-2-[(2S)-2-[(2S)-3-hydroxy-2-[(2S)-2-[(2S)-3-(1H-imidazol-5-yl)-2-{[(2S)-5-oxopyrrolidin-2-yl]formamido}propanamido]-3-(1H-indol-3-yl)propanamido]propanamido]-3-(4-hydroxyphenyl)propanamido]propanamido]-4-methylpentanamido]-5-[(diaminomethylidene)amino]pentanoyl]-N-ethylpyrrolidine-2-carboxamide; acetic acid
Cantidad 10 mg
Peso molecular (g/mol) 1299.50