Learn More
Tocris Bioscience™ BSJ-03-204
Descripción
BSJ-03-204 is a selective Cdk4/6 Degrader (PROTAC™). Displays cereblon-dependent degradation with no effect on IKZF1/3 in Molt4 cells. Induces degradation of Cdk4/6 and G1 cell cycle arrest, and inhibits cell proliferation in a mantle cell lymphoma (MCL) cell line.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[4-[4-[6-[(6-Acetyl-8-cyclopentyl-7,8-dihydro-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]-3-pyridinyl]-1-piperazinyl]butyl]-2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetamide |
| CAS | 2349356-09-6 |
| Cantidad | 5 mg |
| Fórmula molecular | C43H48N10O8 |
| Pureza | 98% |