Learn More
Tocris Bioscience™ BSJ-03-123
Descripción
BSJ-03-123 is a selective Cdk6 Degrader (PROTAC®) (DC50 in sub 10 μM range). Displays no degradation of closely related Cdk4.
Negative control BSJ-Bump (Cat. No. 6922) also available.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| CAS | 2361493-16-3 |
| Cantidad | 5 mg |
| Fórmula molecular | C47H56N10O11 |
| Formula Weight (peso de la fórmula) | 937.02 |
| Tipo de producto | Degrader |
| Pureza | 98% |