Learn More
Tocris Bioscience™ BS 181 dihydrochloride
Descripción
BS 181 dihydrochloride is a selective cdk7 inhibitor (IC50 = 21 nM). Displays >40-fold selectivity for cdk7 over other cdks. Promotes cell cycle arrest and apoptosis in a range of cancer cells in vitro and inhibits MCF-7 tumor xenograft growth in mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N5-(6-Aminohexyl)-3-(1-methylethyl)-N7-(phenylmethyl)pyrazolo[1,5-a]pyrimidine-5,7-diamine dihydrochloride |
| CAS | 1883548-83-1 |
| Cantidad | 50 mg |
| Objetivo | Cyclin-dependent Kinase Inhibitors |
| Fórmula molecular | C22H32N6.2HCl |
| Formula Weight (peso de la fórmula) | 453.45 |
| Pureza | >99% |