Learn More
Maleato de bromfeniramina-d6, TRC
Product Code. 30333141
Change view
Click to view available options
Cantidad:
25 mg
Unit Size:
25 miligramos
Product Code. Cantidad unitSize
30333141 25 mg 25 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30333141 Supplier Toronto Research Chemicals Supplier No. TRCB68635725MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Brompheniramine-d6 Maleate
CAS 1346606-73-2
Fórmula molecular C16 H13 Br D6 N2 . C4 H4 O4
Cantidad 25 mg
Sinónimo gamma-(4-Bromophenyl)-N,N-(dimethyl-d6)-2-pyridinepropanamine Maleate, 2-[p-betaromo-alpha-[2-(dimethylamino-d6)ethyl]benzyl]pyridine Maleate, (+/-)-Brompheniramine-d6 Maleate, Bromphen-d6 Maleate, Dimegan-d6, Dimetane-d6, Parabromdylamine-d6 Maleate, Parabromodylamine-d6 Maleate, Veltane-d6, dl-Brompheniramine-d6 Maleate
Fórmula InChI InChI=1S/C16H19BrN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/i1D3,2D3;
SMILES C(CCN(C([2H])([2H])[2H])C([2H])([2H])[2H])(C1=CC=C(Br)C=C1)C2=CC=CC=N2.OC(=O)\C=C/C(=O)O
Nombre IUPAC 3-(4-bromofenil)-3-piridin-2-il-N,N-bis(trideuteriometil)propan-1-amina; ácido (Z)-but-2-enedioico
Peso molecular (g/mol) 441.349
Formula Weight (peso de la fórmula) 440.122