Learn More
Bragsin2, MedChemExpress
Código de producto. 16407217
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
Tamaño de la unidad:
5 miligramos
16407217 5 mg 5 miligramos
1 options
Código de producto. 16407217
Marca: Medchem Express HY111550
MedChemExpress Bragsin2 is a potent, selective and noncompetitive nucleotide exchange factor BRAG2 inhibitor, with an IC50 of 3 μM. Bragsin2 binds at the interface between the PH domain of BRAG2 and the lipid bilayer, leads BRAG2 unable to activate lipidated Arf GTPase. Bragsin2 affects breast cancer stem cells.
Bragsin2 is a potent, selective and noncompetitive nucleotide exchange factor BRAG2 inhibitor, with an IC50 of 3 μM. Bragsin2 binds at the interface between the PH domain of BRAG2 and the lipid bilayer, leads BRAG2 unable to activate lipidated Arf GTPase. Bragsin2 affects breast cancer stem cells.
Especificaciones
Nombre del producto químico o material Bragsin2
CAS 342795-08-8
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C11H6F3NO5
Cantidad 5 mg
Información de solubilidad DMSO : 100 mg/mL (345.83 mM; Need ultrasonic)
SMILES O=C1C=C(C(F)(F)F)OC2=CC=C(OC)C([N+]([O-])=O)=C21
Peso molecular (g/mol) 289.16
Formula Weight (peso de la fórmula) 289.16
Porcentaje de pureza 99.0%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Cancer-programmed cell death