Learn More
Tocris Bioscience™ BMS 986094
Descripción
BMS 986094 is a potent hepatitis C virus (HCV) replication inhibitor (EC50 = 35 nM). Is rapidly metabolized in primary human hepatocytes to 2'-C-methyl guanosine triphosphate, an inhibitor of HCV RNA-dependent RNA polymerase NS5b. Displays 10-fold reduction in potency in S282T mutant-expressing versus wild-type replicons. Exhibits synergy with Ribavirin (Cat. No. 4501). Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Neopentyl [[[(2 R,3 R,4 R,5 R)-5-(2-amino-6-methoxy-9 H-purin-9-yl)-3,4-dihydroxy-4-methyltetrahydrofuran-2-yl]methoxy](naphthalen-1-yloxy)phosphoryl]-L-alaninate |
| CAS | 1234490-83-5 |
| Cantidad | 5 mg |
| Objetivo | RNA/DNA Polymerase Inhibitors |
| Fórmula molecular | C30H39N6O9P |
| Pureza | 98% |