Learn More
Tocris Bioscience™ BMS 566419
Inosine monophosphatase dehydrogenase (IMPDH) inhibitor
Marca: Tocris Bioscience™ 5492/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BMS 566419 is a potent inosine monophosphate dehydrogenase (IMPDH) inhibitor (IC50 = 17 nM). Inhibits proliferation of human T-lymphoblasts and PBMCs in vitro. Reduces paw swelling in a rat adjuvant arthritis model. Orally available.Especificaciones
| N-[1-[6-(4-Ethyl-1-piperazinyl)-3-pyridinyl]-1-methylethyl]-2-fluoro-9,10-dihydro-9-oxo-3-acridinecarboxamide | |
| 566161-24-8 | |
| 50 mg |