Learn More
BMS 564929, Tocris Bioscience™
Highly potent and muscle-selective androgen receptor (AR) agonist
Marca: Tocris Bioscience 5274/5
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BMS 564929 is a highly potent and selective androgen receptor (AR) agonist (EC50 = 0.44 nM). Exhibits selectivity for ARs over estrogen, glucocorticoid and mineralocorticoid receptors and no significant binding to steroid hormone binding globulin or aromatase. Also exhibits selectivity for muscle over prostate ARs. Active in vivo and orally bioavailable.Especificaciones
| 2-Chloro-3-methyl-4-[(7R,7aS)-tetrahydro-7-hydroxy-1,3-dioxo-1H-pyrrolo[1,2-c]imidazol-2(3H)-yl]benzonitrile | |
| 627530-84-1 | |
| >98% | |
| 5 mg |